Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTN4IP1
Entrez Gene ID
84816
Gene Name
reticulon 4 interacting protein 1
Gene Aliases
NIMP, OPA10
Chromosome Location
Chr.6: 106559237 - 106630921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318746.1 NP_001305675.1 5 - 6 554 84
NM_032730.5 NP_116119.2 5 - 6 1140 84
XM_011536192.2 XP_011534494.1 6 - 7 1228 84
XM_017011375.1 XP_016866864.1 5 - 6 1140 84
XM_017011376.1 XP_016866865.1 5 - 6 1140 84
GenBank mRNA AK095207.1 - 5 - 6 554 84
AK313962.1 - 5 - 6 1058 84
AY063761.1 - 5 - 6 663 84
BC006399.2 - 5 - 6 935 84

View Full Product Details