The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006670.4 | NP_006661.1 | 2 - 3 | 598 | 100 |
XM_011536097.2 | XP_011534399.1 | 1 - 2 | 288 | 100 | |
GenBank mRNA | AK074786.1 | - | 1 - 2 | 84 | 100 |
AK291170.1 | - | 2 - 3 | 598 | 100 | |
BC037161.2 | - | 1 - 2 | 88 | 100 |