Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC45
Entrez Gene ID
8318
Gene Name
cell division cycle 45
Gene Aliases
CDC45L, CDC45L2, PORC-PI-1
Chromosome Location
Chr.22: 19479278 - 19520612 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178010.2 NP_001171481.1 11 - 12 1070 77
NM_001178011.2 NP_001171482.1 9 - 10 836 77
NM_003504.4 NP_003495.1 10 - 11 974 77
XM_005261285.2 XP_005261342.1 12 - 13 1163 77
XM_005261286.3 XP_005261343.1 11 - 12 1067 77
XM_011530415.1 XP_011528717.1 11 - 12 1067 77
XM_011530416.1 XP_011528718.1 10 - 11 870 77
XM_011530417.2 XP_011528719.1 10 - 11 1130 77
XM_011530418.2 XP_011528720.1 10 - 11 1130 77
XM_017028966.1 XP_016884455.1 5 - 6 449 77
XM_017028967.1 XP_016884456.1 2 - 3 189 77
GenBank mRNA AF053074.1 - 10 - 11 855 77
AF062495.1 - 10 - 11 847 77
AF081535.1 - 10 - 11 862 77
AJ223728.1 - 10 - 11 861 77
AK293123.1 - 9 - 10 771 77
AK293338.1 - 11 - 12 967 77
AK302590.1 - 11 - 12 958 77
AK303274.1 - 10 - 11 894 77
AY358971.1 - 10 - 11 897 77
BC006232.2 - 10 - 11 892 77
BC010022.2 - 10 - 11 877 77
BT006792.1 - 10 - 11 831 77
CT841513.1 - 10 - 11 835 77

View Full Product Details