Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LMTK2
Entrez Gene ID
22853
Gene Name
lemur tyrosine kinase 2
Gene Aliases
AATYK2, BREK, KPI-2, KPI2, LMR2, PPP1R100, cprk, hBREK
Chromosome Location
Chr.7: 98106868 - 98209633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB029002.1 - 13 - 14 4617 95

View Full Product Details