The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016094.3 | NP_057178.2 | 3 - 4 | 284 | 140 |
GenBank mRNA | AF059618.1 | - | 3 - 4 | 260 | 140 |
AY542158.1 | - | 3 - 4 | 229 | 140 | |
BC001228.1 | - | 3 - 4 | 263 | 140 | |
BC022206.1 | - | 3 - 4 | 268 | 140 | |
BC046131.1 | - | 3 - 4 | 246 | 140 | |
BC093077.1 | - | 3 - 4 | 274 | 140 | |
BG700015.1 | - | 3 - 4 | 142 | ||
BI792102.1 | - | 3 - 4 | 292 | 140 | |
DA811158.1 | - | 3 - 4 | 283 | 140 |