The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001866.2 | NP_001857.1 | 1 - 2 | 124 | 72 |
GenBank mRNA | AK311879.1 | - | 1 - 2 | 106 | 72 |
BC018386.1 | - | 1 - 2 | 107 | 72 | |
Z14244.1 | - | 1 - 2 | 136 | 72 |