The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003793.3 | NP_003784.2 | 6 - 7 | 956 | 104 |
XM_011545328.2 | XP_011543630.1 | 6 - 7 | 706 | 104 | |
GenBank mRNA | AF071748.1 | - | 4 - 5 | 520 | 104 |
AF071749.1 | - | 4 - 5 | 422 | 104 | |
AF088886.2 | - | 6 - 7 | 956 | 104 | |
AJ007331.1 | - | 6 - 7 | 911 | 104 | |
AK313657.1 | - | 6 - 7 | 924 | 104 | |
AL137742.1 | - | 4 - 5 | 387 | 104 | |
BC011682.2 | - | 6 - 7 | 950 | 104 | |
BC036451.1 | - | 6 - 7 | 931 | 104 | |
CR541680.1 | - | 4 - 5 | 428 | 104 |