Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
Chromosome Location
Chr.11: 66563463 - 66568576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003793.3 NP_003784.2 9 - 10 1257 74
XM_011545328.2 XP_011543630.1 9 - 10 1007 74
GenBank mRNA AF071748.1 - 7 - 8 821 74
AF071749.1 - 7 - 8 723 74
AF088886.2 - 9 - 10 1257 74
AF136279.1 - 9 - 10 1256 74
AJ007331.1 - 9 - 10 1212 74
AK313657.1 - 9 - 10 1225 74
AL137742.1 - 7 - 8 688 74
BC011682.2 - 9 - 10 1251 74
BC036451.1 - 9 - 10 1232 74
CR541680.1 - 8 - 9 729 74

View Full Product Details