The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146256.1 | NP_001139728.1 | 2 - 3 | 706 | 87 |
NM_001146257.1 | NP_001139729.1 | 2 - 3 | 706 | 87 | |
NM_144969.2 | NP_659406.1 | 3 - 4 | 733 | 87 | |
XM_006724624.3 | XP_006724687.1 | 3 - 4 | 270 | 87 | |
XM_017029296.1 | XP_016884785.1 | 3 - 4 | 270 | 87 | |
GenBank mRNA | AK056374.1 | - | 3 - 4 | 271 | 87 |
AK122885.1 | - | 2 - 3 | 455 | 87 | |
AK128587.1 | - | 2 - 3 | 241 | 87 | |
AK303896.1 | - | 3 - 4 | 271 | 87 | |
AY358786.1 | - | 2 - 3 | 627 | 87 | |
BC103980.1 | - | 3 - 4 | 271 | 87 | |
BC103981.1 | - | 3 - 4 | 271 | 87 | |
BC103982.1 | - | 3 - 4 | 271 | 87 |