The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014601.3 | NP_055416.2 | 4 - 5 | 1166 | 77 |
GenBank mRNA | AF181263.1 | - | 3 - 4 | 954 | 77 |
AF454952.1 | - | 3 - 4 | 971 | 77 | |
AK074701.1 | - | 4 - 5 | 1088 | 77 | |
AK097126.1 | - | 4 - 5 | 493 | 77 | |
AK097228.1 | - | 3 - 4 | 485 | 77 | |
AK296907.1 | - | 5 - 6 | 1058 | 77 | |
AK298067.1 | - | 3 - 4 | 629 | 77 | |
AK315548.1 | - | 4 - 5 | 1090 | 77 | |
BC014445.1 | - | 4 - 5 | 1077 | 77 |