The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181741.3 | NP_859525.1 | 1 - 2 | 264 | 89 |
GenBank mRNA | AF022108.1 | - | 1 - 2 | 111 | 89 |
AF047598.1 | - | 1 - 2 | 110 | 89 | |
AF132596.1 | - | 1 - 2 | 97 | 89 | |
AK225092.1 | - | 1 - 2 | 72 | 89 | |
AK299749.1 | - | 1 - 2 | 122 | 89 | |
BC005388.1 | - | 1 - 2 | 76 | 89 | |
BC014847.1 | - | 1 - 2 | 98 | 89 | |
CD242597.1 | - | 1 - 2 | 148 | 89 | |
DA227202.1 | - | 1 - 2 | 264 | 89 |