Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAZAP1
Entrez Gene ID
26528
Gene Name
DAZ associated protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 1407569 - 1435687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018959.3 NP_061832.2 9 - 10 938 114
NM_170711.2 NP_733829.1 9 - 10 938 114
XM_005259531.3 XP_005259588.1 9 - 10 888 114
XM_005259534.3 XP_005259591.1 8 - 9 814 114
XM_005259535.3 XP_005259592.1 8 - 9 813 114
XM_005259536.3 XP_005259593.1 8 - 9 816 114
XM_011527904.2 XP_011526206.1 9 - 10 2203 114
XM_011527905.2 XP_011526207.1 9 - 10 2211 114
XM_011527906.2 XP_011526208.1 9 - 10 2208 114
XM_011527907.2 XP_011526209.1 9 - 10 2217 114
XM_011527908.2 XP_011526210.1 9 - 10 2210 114
XM_011527909.2 XP_011526211.1 9 - 10 2210 114
XM_011527910.2 XP_011526212.1 9 - 10 1290 114
XM_017026581.1 XP_016882070.1 9 - 10 933 114
GenBank mRNA AF181719.1 - 9 - 10 841 114
AK056850.1 - 7 - 8 561 114
AK124583.1 - 2 - 3 2680 114
AK125032.1 - 7 - 8 2151 114
BC012062.1 - 9 - 10 806 114
KU178593.1 - 9 - 10 733 114

View Full Product Details