The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019121.1 | NP_061994.1 | 9 - 10 | 1283 | 65 |
XM_006723159.2 | XP_006723222.1 | 9 - 10 | 1283 | 65 | |
GenBank mRNA | AB075866.1 | - | 2 - 3 | 2960 | 65 |