Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NECAP1
Entrez Gene ID
25977
Gene Name
NECAP endocytosis associated 1
Gene Aliases
EIEE21
Chromosome Location
Chr.12: 8082211 - 8097777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015509.3 NP_056324.2 7 - 8 853 80
NR_024260.1 6 - 7 771 80
GenBank mRNA AK074858.1 - 7 - 8 810 80
AK074880.1 - 7 - 8 823 80
AK074923.1 - 7 - 8 796 80
AK075013.1 - 7 - 8 830 80
AK094805.1 - 2 - 3 1395 80
AK290537.1 - 6 - 7 696 80
AK314424.1 - 6 - 7 705 80
AL050272.1 - 6 - 7 697 80
BC002888.2 - 7 - 8 755 80
BC067367.1 - 5 - 6 525 80
BC084551.1 - 3 - 4 364 80
BC110876.1 - 7 - 8 787 80

View Full Product Details