The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018385.2 | NP_060855.2 | 1 - 2 | 413 | 68 |
GenBank mRNA | AK002163.1 | - | 1 - 2 | 146 | 68 |
AK225094.1 | - | 1 - 2 | 114 | 68 | |
AL136897.1 | - | 1 - 2 | 118 | 68 | |
BC015042.1 | - | 1 - 2 | 118 | 68 | |
BC040119.1 | - | 1 - 2 | 98 | 68 | |
BC058891.1 | - | 1 - 2 | 104 | 68 | |
BC068500.1 | - | 1 - 2 | 127 | 68 | |
DA101432.1 | - | 1 - 2 | 414 | 68 | |
DA414880.1 | - | 1 - 2 | 387 | 68 |