The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040177.2 | NP_001035267.1 | 9 - 10 | 1062 | 121 |
NM_001271021.1 | NP_001257950.1 | 7 - 8 | 891 | 121 | |
NM_001271025.1 | NP_001257954.1 | 6 - 7 | 768 | 121 | |
XM_011519715.2 | XP_011518017.1 | 9 - 10 | 1359 | 121 | |
XM_011519724.2 | XP_011518026.1 | 7 - 8 | 1187 | 121 | |
GenBank mRNA | AB040821.1 | - | 7 - 8 | 772 | 121 |
AB040822.1 | - | 6 - 7 | 649 | 121 | |
AF263242.1 | - | 9 - 10 | 934 | 121 |