Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
Chromosome Location
Chr.2: 187341963 - 187448307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271751.1 NP_001258680.1 11 - 12 1288 86
NM_005795.5 NP_005786.1 12 - 13 1453 86
XM_005246231.3 XP_005246288.2 13 - 14 1497 86
XM_005246232.3 XP_005246289.2 14 - 15 1538 86
XM_005246234.3 XP_005246291.2 12 - 13 1369 86
XM_017003151.1 XP_016858640.1 13 - 14 1428 86
GenBank mRNA AK291630.1 - 12 - 13 1411 86
AK292998.1 - 11 - 12 1246 86
AY389506.1 - 9 - 10 905 86
L76380.1 - 12 - 13 1459 86
U17473.1 - 12 - 13 1375 86

View Full Product Details