Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
Chromosome Location
Chr.2: 187341963 - 187448307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005795.5 NP_005786.1 3 - 4 507 134
XM_005246231.3 XP_005246288.2 4 - 5 551 134
XM_005246232.3 XP_005246289.2 5 - 6 592 134
GenBank mRNA AK291630.1 - 3 - 4 465 134
AL602685.1 - 3 - 4 464 134
DA097876.1 - 3 - 4 465 134
U17473.1 - 3 - 4 429 134

View Full Product Details