The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271751.1 | NP_001258680.1 | 8 - 9 | 1014 | 108 |
NM_005795.5 | NP_005786.1 | 9 - 10 | 1179 | 108 | |
XM_005246231.3 | XP_005246288.2 | 10 - 11 | 1223 | 108 | |
XM_005246232.3 | XP_005246289.2 | 11 - 12 | 1264 | 108 | |
XM_005246234.3 | XP_005246291.2 | 9 - 10 | 1095 | 108 | |
XM_017003151.1 | XP_016858640.1 | 10 - 11 | 1154 | 108 | |
GenBank mRNA | AK291630.1 | - | 9 - 10 | 1137 | 108 |
AK292998.1 | - | 8 - 9 | 972 | 108 | |
AY389506.1 | - | 6 - 7 | 631 | 108 | |
L76380.1 | - | 9 - 10 | 1185 | 108 | |
U17473.1 | - | 9 - 10 | 1101 | 108 |