The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000043.5 | NP_000034.1 | 4 - 5 | 756 | 101 |
NM_001320619.1 | NP_001307548.1 | 4 - 5 | 756 | 101 | |
NM_152871.3 | NP_690610.1 | 4 - 5 | 756 | 101 | |
NM_152872.3 | NP_690611.1 | 4 - 5 | 756 | 101 | |
NR_135314.1 | 4 - 5 | 747 | 101 | ||
XM_006717819.3 | XP_006717882.1 | 5 - 6 | 5438 | 101 | |
XM_011539764.2 | XP_011538066.1 | 5 - 6 | 827 | 101 | |
XM_011539765.2 | XP_011538067.1 | 5 - 6 | 615 | 101 | |
XM_011539766.2 | XP_011538068.1 | 5 - 6 | 567 | 101 | |
XM_011539767.2 | XP_011538069.1 | 4 - 5 | 490 | 101 | |
GenBank mRNA | AB209361.1 | - | 5 - 6 | 4073 | 101 |
AK290978.1 | - | 4 - 5 | 684 | 101 | |
AK311164.1 | - | 4 - 5 | 604 | 101 | |
AK311424.1 | - | 4 - 5 | 747 | 101 | |
AK311499.1 | - | 4 - 5 | 604 | 101 | |
AY495076.2 | - | 4 - 5 | 632 | 101 | |
BC012479.1 | - | 4 - 5 | 748 | 101 | |
CR450307.1 | - | 4 - 5 | 438 | 101 | |
EU481974.2 | - | 4 - 5 | 632 | 101 | |
FJ200481.1 | - | 4 - 5 | 438 | 101 | |
FJ373045.1 | - | 4 - 5 | 438 | 101 | |
FJ373046.1 | - | 4 - 5 | 438 | 101 | |
FM246458.1 | - | 4 - 5 | 438 | 101 | |
FM246459.1 | - | 4 - 5 | 438 | 101 | |
M67454.1 | - | 4 - 5 | 632 | 101 | |
X63717.1 | - | 4 - 5 | 658 | 101 | |
X83493.1 | - | 4 - 5 | 463 | 101 | |
Z47993.1 | - | 4 - 5 | 438 | 101 | |
Z66556.1 | - | 4 - 5 | 438 | 101 |