The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324310.1 | NP_001311239.1 | 5 - 6 | 932 | 65 |
NM_001324311.1 | NP_001311240.1 | 5 - 6 | 942 | 65 | |
NM_014864.3 | NP_055679.1 | 5 - 6 | 1037 | 65 | |
XM_017003001.1 | XP_016858490.1 | 6 - 7 | 983 | 65 | |
XM_017003002.1 | XP_016858491.1 | 6 - 7 | 976 | 65 | |
GenBank mRNA | AB007944.1 | - | 5 - 6 | 1080 | 65 |
AB480690.1 | - | 5 - 6 | 1037 | 65 | |
AK289989.1 | - | 5 - 6 | 937 | 65 | |
BC046441.1 | - | 5 - 6 | 905 | 65 | |
BC051794.1 | - | 5 - 6 | 1037 | 65 |