The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195733.1 | NP_001182662.1 | 15 - 16 | 1873 | 88 |
NM_001300849.1 | NP_001287778.1 | 15 - 16 | 1886 | 88 | |
NM_012398.2 | NP_036530.1 | 15 - 16 | 1873 | 88 | |
XM_011527845.1 | XP_011526147.1 | 15 - 16 | 1880 | 88 | |
XM_011527846.1 | XP_011526148.1 | 14 - 15 | 1847 | 88 | |
XM_011527848.1 | XP_011526150.1 | 15 - 16 | 1880 | 88 | |
XM_011527849.1 | XP_011526151.1 | 15 - 16 | 1880 | 88 | |
XM_011527850.2 | XP_011526152.1 | 15 - 16 | 1839 | 88 | |
XM_011527851.2 | XP_011526153.1 | 8 - 9 | 1467 | 88 | |
XM_011527853.2 | XP_011526155.1 | 8 - 9 | 1298 | 88 | |
XM_017026540.1 | XP_016882029.1 | 14 - 15 | 1847 | 88 | |
GenBank mRNA | AB011161.1 | - | 15 - 16 | 1841 | 88 |
AK301309.1 | - | 15 - 16 | 1856 | 88 | |
AK315912.1 | - | 15 - 16 | 1859 | 88 | |
FJ965536.1 | - | 15 - 16 | 1784 | 88 | |
FJ965537.1 | - | 15 - 16 | 1784 | 88 |