The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018706.6 | NP_061176.3 | 16 - 17 | 2780 | 82 |
GenBank mRNA | BC002477.2 | - | 16 - 17 | 2725 | 82 |
BC007955.2 | - | 16 - 17 | 2707 | 82 | |
CR749726.1 | - | 11 - 12 | 1528 | 82 |