The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018706.6 | NP_061176.3 | 4 - 5 | 839 | 67 |
GenBank mRNA | AK309810.1 | - | 4 - 5 | 747 | 67 |
BC002477.2 | - | 4 - 5 | 784 | 67 | |
BC007955.2 | - | 4 - 5 | 766 | 67 |