Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSX2IP
Entrez Gene ID
117178
Gene Name
SSX family member 2 interacting protein
Gene Aliases
ADIP
Chromosome Location
Chr.1: 84643707 - 84690757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166293.1 NP_001159765.1 12 - 13 1781 84
NM_001166294.1 NP_001159766.1 11 - 12 1649 84
NM_001166295.1 NP_001159767.1 11 - 12 1777 84
NM_001166417.1 NP_001159889.1 13 - 14 1959 84
NM_014021.3 NP_054740.3 13 - 14 1831 84
XM_005270427.1 XP_005270484.1 12 - 13 1720 84
XM_005270428.1 XP_005270485.1 12 - 13 1678 84
XM_005270429.1 XP_005270486.1 12 - 13 1717 84
XM_006710331.1 XP_006710394.1 12 - 13 1638 84
XM_011540610.1 XP_011538912.1 13 - 14 1728 84
XM_017000234.1 XP_016855723.1 12 - 13 1709 84
XM_017000235.1 XP_016855724.1 12 - 13 1684 84
XM_017000236.1 XP_016855725.1 12 - 13 1700 84
XM_017000237.1 XP_016855726.1 11 - 12 1489 84
GenBank mRNA AB023140.1 - 12 - 13 1769 84
AK001710.1 - 8 - 9 991 84
AK093173.1 - 14 - 15 1840 84
AK292475.1 - 12 - 13 1669 84
AK294057.1 - 11 - 12 1536 84
AK307336.1 - 2 - 3 252 84
AK316472.1 - 11 - 12 1555 84
AL133046.1 - 13 - 14 1771 84
AY367055.1 - 13 - 14 1694 84
AY651262.1 - 10 - 11 1351 84
BC033637.1 - 12 - 13 1656 84
BC035580.1 - 11 - 12 1590 84
BC064389.1 - 12 - 13 1742 84

View Full Product Details