The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004261.3 | NP_004252.2 | 4 - 5 | 678 | 111 |
GenBank mRNA | AF051894.1 | - | 4 - 5 | 360 | 111 |
AF288991.1 | - | 4 - 5 | 360 | 111 | |
AK225252.1 | - | 4 - 5 | 369 | 111 | |
AK225640.1 | - | 4 - 5 | 368 | 111 | |
AL833575.1 | - | 4 - 5 | 381 | 111 | |
BC005294.1 | - | 4 - 5 | 386 | 111 | |
BC016359.1 | - | 4 - 5 | 373 | 111 | |
BQ429619.1 | - | 4 - 5 | 393 | 111 |