The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016388.3 | NP_057472.2 | 3 - 4 | 382 | 159 |
GenBank mRNA | AF161547.1 | - | 3 - 4 | 297 | 159 |
AJ224878.1 | - | 3 - 4 | 281 | 159 | |
AK291991.1 | - | 3 - 4 | 382 | 159 | |
BC025713.1 | - | 3 - 4 | 288 | 159 | |
BX415345.2 | - | 3 - 4 | 290 | 159 |