The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006218.3 | NP_006209.2 | 13 - 14 | 2177 | 100 |
XM_006713658.3 | XP_006713721.1 | 13 - 14 | 2202 | 100 | |
XM_011512894.2 | XP_011511196.1 | 13 - 14 | 2283 | 100 | |
GenBank mRNA | AB210020.1 | - | 12 - 13 | 2025 | 100 |
AK292940.1 | - | 13 - 14 | 2135 | 100 | |
AK302608.1 | - | 12 - 13 | 1855 | 100 | |
BC113601.1 | - | 12 - 13 | 2097 | 100 | |
BC113603.1 | - | 12 - 13 | 2097 | 100 | |
U79143.1 | - | 12 - 13 | 2020 | 100 | |
Z29090.1 | - | 12 - 13 | 2032 | 100 |