The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303503.1 | NP_001290432.1 | 8 - 9 | 982 | 54 |
NM_001303504.1 | NP_001290433.1 | 7 - 8 | 874 | 54 | |
NM_001303506.1 | NP_001290435.1 | 8 - 9 | 1035 | 54 | |
NM_001303507.1 | NP_001290436.1 | 7 - 8 | 924 | 54 | |
NM_002720.2 | NP_002711.1 | 8 - 9 | 976 | 54 | |
XM_006721061.3 | XP_006721124.1 | 7 - 8 | 861 | 54 | |
XM_017023403.1 | XP_016878892.1 | 8 - 9 | 952 | 54 | |
XM_017023404.1 | XP_016878893.1 | 7 - 8 | 888 | 54 | |
GenBank mRNA | AF097996.1 | - | 8 - 9 | 898 | 54 |
BC001416.2 | - | 8 - 9 | 929 | 54 | |
BT019746.1 | - | 7 - 8 | 794 | 54 | |
X70218.1 | - | 8 - 9 | 929 | 54 |