The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003884.4 | NP_003875.3 | 8 - 9 | 1723 | 73 |
XM_005265528.4 | XP_005265585.1 | 8 - 9 | 1348 | 73 | |
XM_017007423.1 | XP_016862912.1 | 9 - 10 | 1178 | 73 | |
XM_017007424.1 | XP_016862913.1 | 8 - 9 | 1274 | 73 | |
GenBank mRNA | BC060823.1 | - | 8 - 9 | 1328 | 73 |
BC070075.1 | - | 8 - 9 | 1393 | 73 | |
U57317.2 | - | 8 - 9 | 1735 | 73 |