The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080876.3 | NP_543152.1 | 2 - 3 | 644 | 139 |
GenBank mRNA | AB038770.1 | - | 2 - 3 | 325 | 139 |
AB063186.1 | - | 2 - 3 | 649 | 139 | |
AF486808.1 | - | 2 - 3 | 413 | 139 | |
AK314078.1 | - | 2 - 3 | 454 | 139 | |
BC035000.1 | - | 2 - 3 | 451 | 139 | |
BC093958.1 | - | 2 - 3 | 367 | 139 | |
BC112005.1 | - | 2 - 3 | 379 | 139 |