Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP19
Entrez Gene ID
142679
Gene Name
dual specificity phosphatase 19
Gene Aliases
DUSP17, LMWDSP3, SKRP1, TS-DSP1
Chromosome Location
Chr.2: 183078559 - 183100008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080876.3 NP_543152.1 3 - 4 808 106
NR_135688.1 2 - 3 761 106
GenBank mRNA AB038770.1 - 3 - 4 489 106
AB063186.1 - 3 - 4 813 106
AF486808.1 - 3 - 4 577 106
AK314078.1 - 3 - 4 618 106
BC035000.1 - 3 - 4 615 106
BC093958.1 - 3 - 4 531 106
BC112005.1 - 3 - 4 543 106
BC143337.1 - 2 - 3 485 106

View Full Product Details