Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIWIL3
Entrez Gene ID
440822
Gene Name
piwi like RNA-mediated gene silencing 3
Gene Aliases
HIWI3
Chromosome Location
Chr.22: 24719034 - 24774720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008496.3 NP_001008496.2 16 - 17 2462 79
NM_001255975.1 NP_001242904.1 16 - 17 2435 79
NR_045648.1 17 - 18 2645 79
NR_045649.1 17 - 18 2518 79
GenBank mRNA AB079368.1 - 16 - 17 2458 79
AK302409.1 - 16 - 17 2435 79
AK315806.1 - 16 - 17 2435 79
BC144693.1 - 17 - 18 2275 79
BC144696.1 - 17 - 18 2402 79

View Full Product Details