Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIWIL3
Entrez Gene ID
440822
Gene Name
piwi like RNA-mediated gene silencing 3
Gene Aliases
HIWI3
Chromosome Location
Chr.22: 24719034 - 24774720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008496.3 NP_001008496.2 6 - 7 1118 114
NM_001255975.1 NP_001242904.1 6 - 7 1118 114
NR_045648.1 7 - 8 1328 114
NR_045649.1 7 - 8 1201 114
GenBank mRNA AB079368.1 - 6 - 7 1114 114
AK302409.1 - 6 - 7 1118 114
AK315806.1 - 6 - 7 1118 114
BC144693.1 - 7 - 8 958 114
BC144696.1 - 7 - 8 1085 114

View Full Product Details