The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006332.4 | NP_006323.2 | 2 - 3 | 375 | 79 |
GenBank mRNA | AB049659.1 | - | 2 - 3 | 363 | 79 |
AF097362.1 | - | 2 - 3 | 346 | 79 | |
AK123477.1 | - | 1 - 2 | 1052 | 79 | |
AK291551.1 | - | 2 - 3 | 360 | 79 | |
AL558891.3 | - | 2 - 3 | 375 | 79 | |
BC021136.1 | - | 2 - 3 | 344 | 79 | |
BC031020.1 | - | 2 - 3 | 342 | 79 |