The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006332.4 | NP_006323.2 | 5 - 6 | 693 | 108 |
GenBank mRNA | AF097362.1 | - | 5 - 6 | 664 | 108 |
AI022513.1 | - | 2 - 3 | 108 | ||
AL558891.3 | - | 5 - 6 | 693 | 108 | |
BC021136.1 | - | 5 - 6 | 662 | 108 | |
BC031020.1 | - | 5 - 6 | 660 | 108 |