The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256627.1 | NP_001243556.1 | 10 - 11 | 1367 | 81 |
NM_001256629.1 | NP_001243558.1 | 10 - 11 | 1367 | 81 | |
NM_001256630.1 | NP_001243559.1 | 10 - 11 | 1372 | 81 | |
NM_001282218.1 | NP_001269147.1 | 10 - 11 | 1354 | 81 | |
NM_003957.3 | NP_003948.2 | 10 - 11 | 1367 | 81 | |
NR_046331.1 | 10 - 11 | 1367 | 81 | ||
XM_005253218.3 | XP_005253275.1 | 10 - 11 | 1079 | 81 | |
XM_006718374.2 | XP_006718437.1 | 10 - 11 | 1079 | 81 | |
XM_006718376.2 | XP_006718439.1 | 10 - 11 | 991 | 81 | |
XM_011520461.2 | XP_011518763.1 | 10 - 11 | 991 | 81 | |
XM_011520462.2 | XP_011518764.1 | 10 - 11 | 991 | 81 | |
XM_017018532.1 | XP_016874021.1 | 10 - 11 | 991 | 81 | |
XM_017018533.1 | XP_016874022.1 | 10 - 11 | 1079 | 81 | |
XM_017018534.1 | XP_016874023.1 | 10 - 11 | 991 | 81 | |
XM_017018535.1 | XP_016874024.1 | 10 - 11 | 991 | 81 | |
XM_017018536.1 | XP_016874025.1 | 10 - 11 | 1079 | 81 | |
XM_017018537.1 | XP_016874026.1 | 10 - 11 | 991 | 81 | |
XM_017018538.1 | XP_016874027.1 | 5 - 6 | 536 | 81 | |
GenBank mRNA | AF533876.1 | - | 10 - 11 | 1365 | 81 |
AF533877.1 | - | 10 - 11 | 1365 | 81 | |
AF533878.1 | - | 10 - 11 | 1365 | 81 | |
AF533879.1 | - | 10 - 11 | 1365 | 81 | |
AF533880.1 | - | 10 - 11 | 1365 | 81 | |
AJ006701.1 | - | 8 - 9 | 770 | 81 | |
AK074411.1 | - | 7 - 8 | 490 | 81 | |
AK122851.1 | - | 10 - 11 | 1356 | 81 | |
AK131534.1 | - | 10 - 11 | 1372 | 81 | |
AK297211.1 | - | 6 - 7 | 725 | 81 | |
AK297306.1 | - | 5 - 6 | 496 | 81 | |
AY166857.1 | - | 10 - 11 | 1367 | 81 | |
AY505125.1 | - | 10 - 11 | 981 | 81 |