Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXP1
Entrez Gene ID
27086
Gene Name
forkhead box P1
Gene Aliases
12CC4, HSPC215, MFH, QRF1, hFKH1B
Chromosome Location
Chr.3: 70954714 - 71583989 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006713103.2 XP_006713166.1 1 - 2 152 126

View Full Product Details