The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003883.3 | NP_003874.2 | 3 - 4 | 351 | 74 |
GenBank mRNA | AF005482.1 | - | 1 - 2 | 367 | 74 |
AF039703.1 | - | 3 - 4 | 307 | 74 | |
BC000614.1 | - | 3 - 4 | 340 | 74 | |
BM461585.1 | - | 3 - 4 | 341 | 74 | |
DB456387.1 | - | 3 - 4 | 365 | 74 | |
U66914.1 | - | 3 - 4 | 296 | 74 | |
U75696.1 | - | 2 - 3 | 334 | 74 | |
U75697.1 | - | 3 - 4 | 340 | 74 |