Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS1
Entrez Gene ID
10580
Gene Name
sorbin and SH3 domain containing 1
Gene Aliases
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location
Chr.10: 95311773 - 95561420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034954.1 NP_001030126.1 26 - 27 2707 106
NM_001034955.1 NP_001030127.1 26 - 27 2773 106
NM_001034956.1 NP_001030128.1 23 - 24 2260 106
NM_001034957.1 NP_001030129.1 18 - 19 1825 106
NM_001290294.1 NP_001277223.1 26 - 27 2756 106
NM_001290295.1 NP_001277224.1 21 - 22 2039 106
NM_001290296.1 NP_001277225.1 24 - 25 2617 106
NM_001290297.1 NP_001277226.1 21 - 22 2038 106
NM_001290298.1 NP_001277227.1 22 - 23 2143 106
NM_006434.2 NP_006425.2 17 - 18 1657 106
NM_015385.3 NP_056200.1 21 - 22 2240 106
NM_024991.2 NP_079267.1 22 - 23 2135 106
XM_006717589.1 XP_006717652.1 26 - 27 2757 106
XM_006717593.1 XP_006717656.1 20 - 21 1990 106
XM_011539140.1 XP_011537442.1 28 - 29 3501 106
XM_011539150.1 XP_011537452.1 27 - 28 3426 106
XM_011539155.1 XP_011537457.1 22 - 23 3076 106
XM_011539167.1 XP_011537469.1 21 - 22 2359 106
XM_017015500.1 XP_016870989.1 23 - 24 3006 106
XM_017015501.1 XP_016870990.1 27 - 28 3426 106
XM_017015502.1 XP_016870991.1 26 - 27 3333 106
XM_017015503.1 XP_016870992.1 25 - 26 3258 106
XM_017015504.1 XP_016870993.1 26 - 27 2415 106
XM_017015505.1 XP_016870994.1 24 - 25 3174 106
XM_017015506.1 XP_016870995.1 25 - 26 2340 106
XM_017015507.1 XP_016870996.1 25 - 26 3165 106
XM_017015508.1 XP_016870997.1 24 - 25 2247 106
XM_017015509.1 XP_016870998.1 24 - 25 2269 106
XM_017015510.1 XP_016870999.1 23 - 24 3006 106
XM_017015511.1 XP_016871000.1 23 - 24 2163 106
XM_017015512.1 XP_016871001.1 21 - 22 2074 106
XM_017015513.1 XP_016871002.1 21 - 22 2065 106
XM_017015514.1 XP_016871003.1 21 - 22 1981 106
XM_017015515.1 XP_016871004.1 25 - 26 2616 106
XM_017015516.1 XP_016871005.1 24 - 25 2589 106
XM_017015517.1 XP_016871006.1 24 - 25 2532 106
XM_017015518.1 XP_016871007.1 24 - 25 2545 106
XM_017015519.1 XP_016871008.1 23 - 24 2518 106
XM_017015520.1 XP_016871009.1 23 - 24 2457 106
XM_017015521.1 XP_016871010.1 23 - 24 2470 106
XM_017015522.1 XP_016871011.1 22 - 23 2430 106
XM_017015523.1 XP_016871012.1 22 - 23 2386 106
XM_017015524.1 XP_016871013.1 21 - 22 2359 106
XM_017015525.1 XP_016871014.1 25 - 26 2344 106
XM_017015526.1 XP_016871015.1 24 - 25 2247 106
XM_017015527.1 XP_016871016.1 24 - 25 2269 106
XM_017015528.1 XP_016871017.1 23 - 24 2220 106
XM_017015529.1 XP_016871018.1 23 - 24 2172 106
XM_017015530.1 XP_016871019.1 23 - 24 2163 106
XM_017015531.1 XP_016871020.1 23 - 24 2176 106
XM_017015532.1 XP_016871021.1 22 - 23 2145 106
XM_017015533.1 XP_016871022.1 22 - 23 2149 106
XM_017015534.1 XP_016871023.1 23 - 24 2079 106
XM_017015535.1 XP_016871024.1 22 - 23 2101 106
XM_017015536.1 XP_016871025.1 22 - 23 2092 106
XM_017015537.1 XP_016871026.1 21 - 22 2061 106
XM_017015538.1 XP_016871027.1 22 - 23 2052 106
XM_017015539.1 XP_016871028.1 21 - 22 2074 106
XM_017015540.1 XP_016871029.1 21 - 22 2065 106
GenBank mRNA AB037717.1 - 22 - 23 2067 106
AF136380.1 - 16 - 17 1612 106
AF136381.1 - 23 - 24 2572 106
AF330623.1 - 20 - 21 1993 106
AF330624.1 - 6 - 7 691 106
AF356525.1 - 26 - 27 2718 106
AF356526.1 - 26 - 27 2784 106
AF356527.1 - 23 - 24 2271 106
AJ489942.1 - 21 - 22 2098 106
AK300407.1 - 21 - 22 1992 106
AK304130.1 - 10 - 11 1080 106
AL117472.1 - 21 - 22 2198 106
AM260536.1 - 26 - 27 2714 106
BC042612.1 - 21 - 22 2539 106
BC152463.1 - 22 - 23 2067 106
BX640814.1 - 10 - 11 2070 106

View Full Product Details