Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS1
Entrez Gene ID
10580
Gene Name
sorbin and SH3 domain containing 1
Gene Aliases
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location
Chr.10: 95311773 - 95561420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034954.1 NP_001030126.1 1 - 2 56 60
NM_001034955.1 NP_001030127.1 1 - 2 56 60
NM_001034956.1 NP_001030128.1 1 - 2 56 60
NM_001034957.1 NP_001030129.1 1 - 2 56 60
NM_001290294.1 NP_001277223.1 3 - 4 243 60
NM_001290295.1 NP_001277224.1 3 - 4 243 60
NM_001290296.1 NP_001277225.1 1 - 2 56 60
NM_001290297.1 NP_001277226.1 1 - 2 56 60
NM_001290298.1 NP_001277227.1 1 - 2 56 60
NM_006434.2 NP_006425.2 1 - 2 56 60
NM_015385.3 NP_056200.1 3 - 4 243 60
NM_024991.2 NP_079267.1 3 - 4 243 60
XM_006717589.1 XP_006717652.1 3 - 4 196 60
XM_006717593.1 XP_006717656.1 3 - 4 221 60
XM_011539140.1 XP_011537442.1 3 - 4 196 60
XM_011539150.1 XP_011537452.1 3 - 4 196 60
XM_011539155.1 XP_011537457.1 3 - 4 221 60
XM_011539167.1 XP_011537469.1 3 - 4 221 60
XM_017015500.1 XP_016870989.1 3 - 4 196 60
XM_017015501.1 XP_016870990.1 3 - 4 196 60
XM_017015502.1 XP_016870991.1 3 - 4 196 60
XM_017015503.1 XP_016870992.1 3 - 4 196 60
XM_017015504.1 XP_016870993.1 3 - 4 196 60
XM_017015505.1 XP_016870994.1 3 - 4 196 60
XM_017015506.1 XP_016870995.1 3 - 4 196 60
XM_017015507.1 XP_016870996.1 3 - 4 196 60
XM_017015508.1 XP_016870997.1 3 - 4 196 60
XM_017015509.1 XP_016870998.1 3 - 4 221 60
XM_017015510.1 XP_016870999.1 3 - 4 196 60
XM_017015511.1 XP_016871000.1 3 - 4 196 60
XM_017015512.1 XP_016871001.1 3 - 4 221 60
XM_017015513.1 XP_016871002.1 3 - 4 221 60
XM_017015514.1 XP_016871003.1 3 - 4 221 60
XM_017015515.1 XP_016871004.1 3 - 4 196 60
XM_017015516.1 XP_016871005.1 3 - 4 196 60
XM_017015517.1 XP_016871006.1 3 - 4 196 60
XM_017015518.1 XP_016871007.1 3 - 4 221 60
XM_017015519.1 XP_016871008.1 3 - 4 221 60
XM_017015520.1 XP_016871009.1 3 - 4 196 60
XM_017015521.1 XP_016871010.1 3 - 4 221 60
XM_017015522.1 XP_016871011.1 3 - 4 196 60
XM_017015523.1 XP_016871012.1 3 - 4 221 60
XM_017015524.1 XP_016871013.1 3 - 4 221 60
XM_017015525.1 XP_016871014.1 3 - 4 221 60
XM_017015526.1 XP_016871015.1 3 - 4 196 60
XM_017015527.1 XP_016871016.1 3 - 4 221 60
XM_017015528.1 XP_016871017.1 3 - 4 196 60
XM_017015529.1 XP_016871018.1 3 - 4 196 60
XM_017015530.1 XP_016871019.1 3 - 4 196 60
XM_017015531.1 XP_016871020.1 3 - 4 221 60
XM_017015532.1 XP_016871021.1 3 - 4 196 60
XM_017015533.1 XP_016871022.1 3 - 4 221 60
XM_017015534.1 XP_016871023.1 3 - 4 196 60
XM_017015535.1 XP_016871024.1 3 - 4 221 60
XM_017015536.1 XP_016871025.1 3 - 4 221 60
XM_017015537.1 XP_016871026.1 3 - 4 196 60
XM_017015538.1 XP_016871027.1 3 - 4 196 60
XM_017015539.1 XP_016871028.1 3 - 4 221 60
XM_017015540.1 XP_016871029.1 3 - 4 221 60
GenBank mRNA AB037717.1 - 3 - 4 175 60
AF356525.1 - 1 - 2 67 60
AF356526.1 - 1 - 2 67 60
AF356527.1 - 1 - 2 67 60
AK022468.1 - 4 - 5 318 60
AK300407.1 - 3 - 4 196 60
AK304753.1 - 3 - 4 196 60
AL117472.1 - 3 - 4 201 60
AL707894.1 - 4 - 5 359 60
BC042612.1 - 2 - 3 294 60
BC152463.1 - 3 - 4 175 60
DC334283.1 - 3 - 4 244 60

View Full Product Details