Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORBS1
Entrez Gene ID
10580
Gene Name
sorbin and SH3 domain containing 1
Gene Aliases
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location
Chr.10: 95311773 - 95561420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034954.1 NP_001030126.1 6 - 7 486 55
NM_001034955.1 NP_001030127.1 6 - 7 486 55
NM_001290294.1 NP_001277223.1 8 - 9 673 55
NM_001290296.1 NP_001277225.1 5 - 6 459 55
NM_015385.3 NP_056200.1 7 - 8 577 55
XM_006717589.1 XP_006717652.1 7 - 8 599 55
XM_011539167.1 XP_011537469.1 6 - 7 528 55
XM_017015515.1 XP_016871004.1 8 - 9 626 55
XM_017015516.1 XP_016871005.1 7 - 8 599 55
XM_017015517.1 XP_016871006.1 8 - 9 626 55
XM_017015518.1 XP_016871007.1 7 - 8 555 55
XM_017015519.1 XP_016871008.1 6 - 7 528 55
XM_017015520.1 XP_016871009.1 8 - 9 626 55
XM_017015521.1 XP_016871010.1 7 - 8 555 55
XM_017015522.1 XP_016871011.1 7 - 8 599 55
XM_017015523.1 XP_016871012.1 7 - 8 555 55
XM_017015524.1 XP_016871013.1 6 - 7 528 55
GenBank mRNA AF136381.1 - 4 - 5 414 55
AF356525.1 - 6 - 7 497 55
AF356526.1 - 6 - 7 497 55
AK022468.1 - 8 - 9 652 55
AL117472.1 - 7 - 8 535 55
AM260536.1 - 8 - 9 631 55
BC042612.1 - 5 - 6 601 55

View Full Product Details