The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016013.3 | NP_057097.2 | 2 - 3 | 971 | 75 |
NR_045620.1 | 2 - 3 | 971 | 75 | ||
XM_006720555.2 | XP_006720618.1 | 2 - 3 | 901 | 75 | |
XM_011521658.1 | XP_011519960.1 | 2 - 3 | 835 | 75 | |
XM_011521659.2 | XP_011519961.1 | 2 - 3 | 998 | 75 | |
XM_017022293.1 | XP_016877782.1 | 2 - 3 | 902 | 75 | |
GenBank mRNA | AF151823.1 | - | 2 - 3 | 75 | |
AK309889.1 | - | 2 - 3 | 971 | 75 | |
AK310983.1 | - | 2 - 3 | 971 | 75 | |
BC000780.2 | - | 2 - 3 | 955 | 75 |