Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFAF1
Entrez Gene ID
51103
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 1
Gene Aliases
CGI-65, CGI65, CIA30
Chromosome Location
Chr.15: 41387349 - 41402694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016013.3 NP_057097.2 4 - 5 1231 69
NR_045620.1 5 - 6 1485 69
XM_006720555.2 XP_006720618.1 4 - 5 1161 69
XM_011521658.1 XP_011519960.1 4 - 5 1095 69
GenBank mRNA AF151823.1 - 4 - 5 1194 69
AI911511.1 - 1 - 2 69
AK310983.1 - 5 - 6 1485 69
BC000780.2 - 4 - 5 1215 69
BG192055.1 - 2 - 3 263 69

View Full Product Details