Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZG16B
Entrez Gene ID
124220
Gene Name
zymogen granule protein 16B
Gene Aliases
EECP, HRPE773, JCLN2, PAUF, PRO1567
Chromosome Location
Chr.16: 2830172 - 2832284 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145252.2 NP_660295.2 2 - 3 232 105
GenBank mRNA AK311810.1 - 2 - 3 100 105
AY359021.1 - 2 - 3 174 105
BC009722.1 - 2 - 3 172 105
BQ924772.1 - 2 - 3 235 105
EF067313.1 - 2 - 3 131 105
JN411265.1 - 2 - 3 173 105

View Full Product Details