Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL2
Entrez Gene ID
54843
Gene Name
synaptotagmin like 2
Gene Aliases
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location
Chr.11: 85694221 - 85811182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162952.2 NP_001156424.1 1 - 2 565 71
NM_001289608.1 NP_001276537.1 9 - 10 1724 71
NM_001289609.1 NP_001276538.1 1 - 2 565 71
NM_001289610.1 NP_001276539.1 2 - 3 249 71
NM_032943.4 NP_116561.1 10 - 11 2214 71
NM_206929.3 NP_996812.1 2 - 3 249 71
XM_005274060.1 XP_005274117.1 9 - 10 1738 71
XM_005274061.1 XP_005274118.1 9 - 10 1735 71
XM_005274062.2 XP_005274119.1 10 - 11 2159 71
XM_005274063.1 XP_005274120.1 9 - 10 1738 71
XM_011545105.1 XP_011543407.1 9 - 10 5602 71
XM_011545106.1 XP_011543408.1 9 - 10 5599 71
XM_011545107.2 XP_011543409.1 10 - 11 6023 71
XM_011545108.1 XP_011543410.1 9 - 10 5602 71
XM_011545109.2 XP_011543411.1 9 - 10 5584 71
XM_011545110.1 XP_011543412.1 9 - 10 5602 71
XM_017017934.1 XP_016873423.1 10 - 11 6020 71
XM_017017936.1 XP_016873425.1 6 - 7 5096 71
XM_017017937.1 XP_016873426.1 9 - 10 1639 71
XM_017017938.1 XP_016873427.1 9 - 10 1717 71
XM_017017939.1 XP_016873428.1 2 - 3 1905 71
GenBank mRNA AB046817.1 - 10 - 11 2225 71
AK131365.1 - 3 - 4 842 71
AK296224.1 - 9 - 10 1706 71
AK298604.1 - 2 - 3 167 71
AL601023.1 - 2 - 3 222 71
BC068495.1 - 1 - 2 290 71
BC110315.1 - 1 - 2 94 71
BX647671.1 - 3 - 4 2545 71
CB046277.1 - 2 - 3 206 71
DA378461.1 - 2 - 3 249 71

View Full Product Details