Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL2
Entrez Gene ID
54843
Gene Name
synaptotagmin like 2
Gene Aliases
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location
Chr.11: 85694221 - 85811182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162951.2 NP_001156423.1 6 - 7 1732 108
NM_001162953.2 NP_001156425.1 6 - 7 1735 108
NM_001289608.1 NP_001276537.1 6 - 7 1493 108
NM_032943.4 NP_116561.1 7 - 8 1983 108
XM_005274057.1 XP_005274114.1 6 - 7 1507 108
XM_005274060.1 XP_005274117.1 6 - 7 1507 108
XM_005274061.1 XP_005274118.1 6 - 7 1504 108
XM_005274062.2 XP_005274119.1 7 - 8 1928 108
XM_005274063.1 XP_005274120.1 6 - 7 1507 108
XM_005274066.1 XP_005274123.1 6 - 7 1504 108
XM_011545105.1 XP_011543407.1 6 - 7 1507 108
XM_011545106.1 XP_011543408.1 6 - 7 1504 108
XM_011545107.2 XP_011543409.1 7 - 8 1928 108
XM_011545108.1 XP_011543410.1 6 - 7 1507 108
XM_011545109.2 XP_011543411.1 6 - 7 1489 108
XM_011545110.1 XP_011543412.1 6 - 7 1507 108
XM_011545112.1 XP_011543414.1 6 - 7 1507 108
XM_017017933.1 XP_016873422.1 6 - 7 1504 108
XM_017017934.1 XP_016873423.1 7 - 8 1925 108
XM_017017935.1 XP_016873424.1 6 - 7 1507 108
XM_017017936.1 XP_016873425.1 3 - 4 1001 108
XM_017017937.1 XP_016873426.1 6 - 7 1408 108
XM_017017938.1 XP_016873427.1 6 - 7 1486 108
GenBank mRNA AB046817.1 - 7 - 8 1994 108
AK296224.1 - 6 - 7 1475 108
AK296740.1 - 5 - 6 1591 108
BC136450.1 - 6 - 7 1667 108
BC144114.1 - 6 - 7 1667 108
BC144115.1 - 6 - 7 1664 108
EU035829.1 - 6 - 7 1388 108

View Full Product Details