Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEPT10
Entrez Gene ID
151011
Gene Name
septin 10
Gene Aliases
-
Chromosome Location
Chr.2: 109542797 - 109614247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321496.1 NP_001308425.1 7 - 8 1227 83
NM_001321498.1 NP_001308427.1 9 - 10 1541 83
NM_001321499.1 NP_001308428.1 6 - 7 1040 83
NM_001321500.1 NP_001308429.1 8 - 9 1472 83
NM_001321501.1 NP_001308430.1 9 - 10 1545 83
NM_001321504.1 NP_001308433.1 6 - 7 1158 83
NM_001321505.1 NP_001308434.1 6 - 7 1158 83
NM_001321506.1 NP_001308435.1 5 - 6 971 83
NM_001321507.1 NP_001308436.1 7 - 8 1227 83
NM_001321509.1 NP_001308438.1 9 - 10 1545 83
NM_001321510.1 NP_001308439.1 6 - 7 1040 83
NM_001321511.1 NP_001308440.1 5 - 6 971 83
NM_001321512.1 NP_001308441.1 9 - 10 1541 83
NM_001321513.1 NP_001308442.1 9 - 10 1678 83
NM_001321514.1 NP_001308443.1 9 - 10 1369 83
NM_001321515.1 NP_001308444.1 8 - 9 1609 83
NM_144710.4 NP_653311.1 9 - 10 1541 83
NM_178584.3 NP_848699.1 8 - 9 1472 83
XM_006712317.1 XP_006712380.1 9 - 10 1541 83
XM_011510698.2 XP_011509000.1 9 - 10 1374 83
XM_011510699.2 XP_011509001.1 8 - 9 1193 83
XM_011510700.2 XP_011509002.1 9 - 10 1544 83
XM_011510701.2 XP_011509003.1 7 - 8 1268 83
XM_011510702.2 XP_011509004.1 6 - 7 1039 83
XM_011510703.2 XP_011509005.1 9 - 10 1151 83
GenBank mRNA AB208875.1 - 5 - 6 635 83
AF146760.1 - 9 - 10 1288 83
AK021681.1 - 9 - 10 1541 83
AK022682.1 - 6 - 7 873 83
AK092033.1 - 8 - 9 1192 83
AK292038.1 - 9 - 10 1259 83
AK294427.1 - 6 - 7 927 83
AK295560.1 - 7 - 8 937 83
AL049934.1 - 3 - 4 344 83
BC020502.1 - 9 - 10 1235 83
BC050345.1 - 9 - 10 1478 83

View Full Product Details