The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003854.2 | NP_003845.2 | 3 - 4 | 422 | 133 |
XM_005264035.1 | XP_005264092.1 | 3 - 4 | 415 | 133 | |
XM_005264036.1 | XP_005264093.1 | 3 - 4 | 337 | 133 | |
XM_011512091.1 | XP_011510393.1 | 3 - 4 | 415 | 133 | |
XM_011512092.1 | XP_011510394.1 | 3 - 4 | 415 | 133 | |
XM_011512093.1 | XP_011510395.1 | 3 - 4 | 415 | 133 | |
XM_011512094.1 | XP_011510396.1 | 3 - 4 | 337 | 133 | |
XM_011512096.1 | XP_011510398.1 | 3 - 4 | 415 | 133 | |
XM_017005174.1 | XP_016860663.1 | 3 - 4 | 415 | 133 | |
GenBank mRNA | AF284434.1 | - | 3 - 4 | 430 | 133 |
AK315334.1 | - | 3 - 4 | 337 | 133 | |
U49065.1 | - | 3 - 4 | 430 | 133 |