Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL1RL2
Entrez Gene ID
8808
Gene Name
interleukin 1 receptor like 2
Gene Aliases
IL-1Rrp2, IL-36R, IL1R-rp2, IL1RRP2
Chromosome Location
Chr.2: 102186973 - 102242906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003854.2 NP_003845.2 4 - 5 606 70
XM_005264035.1 XP_005264092.1 4 - 5 599 70
XM_005264036.1 XP_005264093.1 4 - 5 521 70
XM_011512091.1 XP_011510393.1 4 - 5 599 70
XM_011512092.1 XP_011510394.1 4 - 5 599 70
XM_011512093.1 XP_011510395.1 4 - 5 599 70
XM_011512094.1 XP_011510396.1 4 - 5 521 70
XM_011512096.1 XP_011510398.1 4 - 5 599 70
XM_017005174.1 XP_016860663.1 4 - 5 599 70
GenBank mRNA AF284434.1 - 4 - 5 614 70
AK315334.1 - 4 - 5 521 70
U49065.1 - 4 - 5 614 70

View Full Product Details